missing translation for 'onlineSavingsMsg'
Learn More

Lysozyme Antibody (LYZ/3944), DyLight 650, Novus Biologicals™

Product Code. 18369913 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369913 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369913 Supplier Novus Biologicals Supplier No. NBP314115C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lysozyme Monoclonal antibody specifically detects Lysozyme in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lysozyme
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LYZ/3944
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis
Host Species Mouse
Immunogen Recombinant fragment of human Lysozyme protein (around aa 18-147) (exact sequence is proprietary) (Uniprot: P61626)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4069
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.