missing translation for 'onlineSavingsMsg'
Learn More

Lysosomal Pro-X Carboxypeptidase/PRCP Antibody [DyLight 550], Novus Biologicals™

Product Code. 30737204 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737204 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737204 Supplier Novus Biologicals Supplier No. AF7164L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Lysosomal Pro-X Carboxypeptidase/PRCP Polyclonal antibody specifically detects Lysosomal Pro-X Carboxypeptidase/PRCP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lysosomal Pro-X Carboxypeptidase/PRCP
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Angiotensinase C, EC 3.4.16.2, HUMPCP, Lysosomal carboxypeptidase C, lysosomal Pro-X carboxypeptidase, PCPMGC2202, Proline carboxypeptidase, Prolylcarboxypeptidase, prolylcarboxypeptidase (angiotensinase C), prolylcarboxypeptidase isoform 1 preproprotein
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Lysosomal Pro-X Carboxypeptidase/PRCP, Met1-His496, Accession # P42785
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5547
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.