missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYSMD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69234
This item is not returnable.
View return policy
Description
LYSMD4 Polyclonal specifically detects LYSMD4 in Human samples. It is validated for Western Blot.
Specifications
| LYSMD4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ33008, lysM and putative peptidoglycan-binding domain-containing protein 4, LysM, putative peptidoglycan-binding, domain containing 4, MGC99501 | |
| Rabbit | |
| 32 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| A6NII6 | |
| LYSMD4 | |
| Synthetic peptides corresponding to LYSMD4(LysM, putative peptidoglycan-binding, domain containing 4) The peptide sequence was selected from the N terminal of LYSMD4. Peptide sequence PRREQVTWCCCSGSWPRRTASTSWRCSMAANTFYFRPNGAGDTRQNLIPD. | |
| Affinity purified | |
| RUO | |
| 145748 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction