Learn More
LYSMD3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24313-1-AP-20UL
This item is not returnable.
View return policy
Description
The LYSMD3 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human LYSMD3. This antibody recognizes human, mouse antigen. The LYSMD3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| LYSMD3 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 116068 | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 440 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |