missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LYSMD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553706
This item is not returnable.
View return policy
Description
Immunogen sequence: FSSLTETLCP PKGRQTSRHS SVQYSSEQQE ILPANDSLAY SDSAGSFLKE VDRDIEQIVK CTDNKRENLN EVVSALTAQQ MRFEPDNKNT QRKDPYYGAD W Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 75%.
LYSMD3 is a protein coding gene.
Specifications
| LYSMD3 | |
| Polyclonal | |
| Unconjugated | |
| LYSMD3 | |
| 1110030H10Rik; BC003322; lysM and putative peptidoglycan-binding domain-containing protein 3; LysM domain containing 3; LysM, putative peptidoglycan-binding, domain containing 3; LYSMD3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116068 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z3D4 | |
| LYSMD3 | |
| Recombinant protein corresponding to Human LYSMD3. Recombinant protein control fragment (Product #RP-90483). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction