missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Lysis Buffer VL

Product Code. 15889117
Click to view available options
Quantity:
200 mL
Unit Size:
200mL
This item is not returnable. View return policy

Product Code. 15889117

Brand: Macherey-Nagel™ 740833.200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lysis Buffer VL to be used with NucleoSpin Virus or NucleoSnap cfDNA kits

Specifications

Chemical Name or Material Lysis buffer VL
CAS 50-01-1
Color Colorless
Concentration Guanidine hydrochloride, Water
For Use With (Application) Purification of DNA
For Use With (Equipment) Compatible kits (NucleoSnap DNA Plasma, NucleoSpin Virus)
pH 4.5 to 5.2
Physical Form Liquid
Quantity 200 mL
Formulation Guanidine hydrochloride, Water
Odor Odorless
Show More Show Less
compliance-icons
Product Identifier
  • Buffer VL
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash with water thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.