Learn More
Macherey-Nagel™ Lysis Buffer S2-EF
Product Code. 30694916
Change view
Click to view available options
Quantity:
3 L
Unit Size:
3 L
Product Code. Quantity unitSize
30694916 3 L 3 L
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30694916 Supplier Macherey-Nagel™ Supplier No. 740791.3
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Lysis Buffer S2‐EF to be used with NucleoBond PC EF kits
  • For NucleoBond PC EF kits
Specifications
For Use With (Application) Endotoxin-free plasmid DNA isolation
Quantity 3 L
Solution Type Buffer