missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Lysis Buffer P
Description
- For use with NucleoType Seed PCR Kit
- Enables a simple sample preparation
- The lysis procedure releases DNA in sufficient amount and quality to serve as template for the subsequent PCR
- The seed sample is placed into an adequate volume of Lysis Buffer P containing Proteinase K
Specifications
Specifications
| Chemical Name or Material | Lysis Buffer P |
| For Use With (Equipment) | NucleoType Seed PCR Kit |
| Physical Form | Separate Lysis Buffer |
| Quantity | 250 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?