missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Lysis Buffer DL

Product Code. 15800898
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15800898 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15800898 Supplier Macherey-Nagel™ Supplier No. 740202.32
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lysis Buffer DL to be used with NucleoSpin RNA Blood kits

The NucleoSpin™ 8 RNA Blood kit offers a direct total blood lysis from up to 400 μL whole blood collected in standard blood collection tubes and prevents RNA degradation during the isolation procedure.

  • For use with NucleoSpin™ 8 RNA Blood kit
  • Solution containing large amounts of chaotropic ions
  • Along with Buffer RB4, creates binding conditions for adsorption of RNA to the silica membrane
  • Selective erythrocyte lysis and preparation of a leukocyte pellet is not necessary
  • Sufficient for 24 x 8 preps

Specifications

CAS 593-84-0
Color Colorless
For Use With (Application) RNA Isolation
For Use With (Equipment) NucleoSpin™ 8 Blood kit (MPN 740220)
Physical Form Solution
Quantity 100 mL
Formulation Guanidinium thiocyanate
compliance-icons
Product Identifier
  • Buffer DL
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.