missing translation for 'onlineSavingsMsg'
Learn More

Lysipressin Acetic Acid Salt, TRC

Product Code. 30483618
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483618 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483618 Supplier Toronto Research Chemicals Supplier No. TRCL4887705MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 1116.3
Quantity 5 mg
Shipping Condition Room Temperature
Synonym 19-Amino-4-[2-[[5-amino-1-[(carbamoylmethyl)carbamoyl]pentyl]carbamoyl]-1-pyrrolidinylcarbonyl]-13-benzyl-7-(carbamoylmethyl)-16-p-hydroxybenzyl-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentaazacycloeicosane-10-propionamide Acetic Acid Salt∣1,2-Dithia-5,8,11,14,17-pentaazacycloeicosane, cyclic peptide deriv. Acetic Acid Salt∣3-(Phenylalanine)-8-lysine oxytocin∣8-L-Lysine vasopressin∣Diapid∣Glycinamide, L-cysteinyl-L-tyrosyl-L-phenylalanyl-L-glutaminyl-L-asparaginyl-L-cysteinyl-L-prolyl-L-lysyl-, cyclic (1-6)-disulfide Acetic Acid Salt∣L 8∣L-Lysine vasopressin Acetic Acid Salt∣Lypressin Acetic Acid Salt∣Lysine pitressin Acetic Acid Salt∣Lysine vasopressin Acetic Acid Salt∣Lysine-ADH Acetic Acid Salt∣Lysopressin Acetic Acid Salt∣Lysylvasopressin Acetic Acid Salt∣
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.