missing translation for 'onlineSavingsMsg'
Learn More

Lysino Norleucine, TRC

Product Code. 30012461
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012461 1 mg 1mg
30009894 2.5 mg 2.5mg
30006402 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012461 Supplier Toronto Research Chemicals Supplier No. TRCL4887501MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25612-46-8
Molecular Formula C12 H25 N3 O4
Molecular Weight (g/mol) 275.34
IUPAC Name (2S)-2-amino-6-[[(5S)-5-amino-5-carboxypentyl]amino]hexanoic acid
SMILES N[C@@H](CCCCNCCCC[C@H](N)C(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C12H25N3O4/c13-9(11(16)17)5-1-3-7-15-8-4-2-6-10(14)12(18)19/h9-10,15H,1-8,13-14H2,(H,16,17)(H,18,19)/t9-,10-/m0/s1
Quantity 1 mg
Formula Weight 275.1845
Shipping Condition Room Temperature
Chemical Name or Material Lysinonorleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.