missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix S, 96-Tube Rack
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
- Product Overview: Each impact-resistant 1.2 mL well contains four (4) 1/8 in. (3.175 mm) diameter stainless steel beads. Use to break hard/brittle samples. Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
- Applications: Metal Bead Lysing Matrix is used to break hard/brittle samples. Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
Specifications
Specifications
| DNase | Dnase-free |
| For Use With (Equipment) | FastPrep-96 |
| For Use With (Application) | Sample Preparation, Sample lysis, Nucleic Acid isolation and purification, Molecular biology, Gene expression, Microbiological analysis, Gene function, Gene regulation |
| Matrix | Lysing Matrix S |
| Quantity | 1 Ea. |
| RNase | Rnase-free |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction