missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix B, 96-well Rack

Product Code. 15113063 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 Ea.
10 Pk.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15113063 10 Pk. Each
15212870 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15113063 Supplier MP Biomedicals™ Supplier No. 116981010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lysing Matrix B, 96-tube Rack is pre-filled with 0.1 mm silica spheres and used to lyse bacteria and spores.

Lysing Matrix B, 96-tube rack comes with 1.2 mL cluster-strip tube and cap assemblies pre-filled with 0.1 mm silica spheres. Use with FastPrep-96 instrument or similar bead beater.

  • Tube size: 96-tube rack
  • Sample types: Gram positive and negative bacteria, fungal tissues, spores
  • Matrix composition: 0.1 mm silica spheres
TRUSTED_SUSTAINABILITY

Specifications

Bead Type Silica Spheres
Content And Storage Store at Room Temperature.
Sample Type Bacteria, Fungi, Soil, Animal Tissues
DNase Dnase-free
For Use With (Equipment) FastPrep-96 or other bead beaters
For Use With (Application) Nucleic Acid Isolation and Purification, Homogenization
Includes 1.2 mL Cluster-strip Tube and Cap Assemblies
Matrix Lysing Matrix B
Quantity 10 Pk.
RNase RNase-free
Sample Volume Less than 80 mg per prep
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.