missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06855SS
This item is not returnable.
View return policy
Description
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2 to 5 μg/mL, Immunohistochemistry-Paraffin 1:200 | |
| EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B | |
| Partial recombinant protein made to an internal portion of the human JMJD3 protein (amino acids 1520-1682) [UniProt O15054] | |
| 0.025 mg | |
| Cancer, Chromatin Research, Transcription Factors and Regulators | |
| 23135 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction