missing translation for 'onlineSavingsMsg'
Learn More

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-92982-0.02ml

 View more versions of this product

Product Code. 18676160

  • £153.00 / 0.02mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3
Polyclonal
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B
Recombinant fusion protein containing a sequence corresponding to amino acids 90-190 of mouse Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 (NP_001017426.1). LESLHGCVQALLREPAQPGLWEQLGQLYESEHDSEEAVCCYHRALRYGGSFAELGPRIGRLQQAQLWNFHAGSCQHRAKVLPPLEQVWNLLHLEHKRNYGA
0.02 mL
Cancer, Chromatin Research, Transcription Factors and Regulators
23135
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunofluorescence
Unconjugated
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.