missing translation for 'onlineSavingsMsg'
Learn More

Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Antibody (OTI5B9), HRP, Novus Biologicals™

Product Code. 18103864 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18103864 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18103864 Supplier Novus Biologicals Supplier No. NBP272553H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Monoclonal specifically detects Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5B9
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias EC 1.14.11, EC 1.14.11.-, GASC-1 protein, GASC1JmjC domain-containing histone demethylation protein 3C, Gene amplified in squamous cell carcinoma 1 protein, JHDM3C, JMJD2CbA146B14.1, jumonji domain containing 2C, Jumonji domain-containing protein 2C, KIAA0780FLJ25949, lysine (K)-specific demethylase 4C, lysine-specific demethylase 4C
Gene Symbols KDM4C
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Chromatin Modifiers, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23081.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.