missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49602SS
This item is not returnable.
View return policy
Description
Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Polyclonal specifically detects Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
| Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| EC 1.14.11, EC 1.14.11.-, JHDM3AJMJD2, JmjC domain-containing histone demethylation protein 3A, JMJD2Alysine-specific demethylase 4A, jumonji domain containing 2, jumonji domain containing 2A, Jumonji domain-containing protein 2A, KIAA0677jumonji C domain-containing histone demethylase 3A, lysine (K)-specific demethylase 4A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.53 mg/mL | |
| Western Blot 1:25000, Simple Western 1:500, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| O75164 | |
| KDM4A | |
| A genomic peptide made to an internal region of the human JMJD2A protein (within residues 320-460). [Swiss-Prot O75164] | |
| 0.025 mL | |
| Chromatin Modifiers, Epigenetics | |
| 9682 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction