missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Antibody (1E12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£187.00 - £381.00
Specifications
| Antigen | Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A |
|---|---|
| Clone | 1000000000000 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Simple Western 1:500, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261585
|
Novus Biologicals
NBP1-51678SS |
0.025 mL |
£187.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18242261
|
Novus Biologicals
NBP1-51678 |
0.1 mL |
£381.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A Monoclonal specifically detects Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A in Human samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Chromatin Modifiers, Epigenetics | |
| Q9Y4C1 | |
| 55818 | |
| Purified recombinant fragment of human JMJD1A expressed in E. coli. [UniProt# Q9Y4C1] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1000000000000 | |
| Western Blot 1:500-1:2000, Simple Western 1:500, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| Human | |
| EC 1.14.11, EC 1.14.11.-, JHDM2AJMJD1AJHMD2A, JmjC domain-containing histone demethylation protein 2A, JMJD1, jumonji C domain-containing histone demethylase 2A, jumonji domain containing 1, jumonji domain containing 1A, Jumonji domain-containing protein 1A, KIAA0742DKFZp686A24246, lysine (K)-specific demethylase 3A, lysine-specific demethylase 3A, testis-specific protein A, TSGADKFZp686P07111 | |
| KDM3A | |
| IgG1 | |
| Ammonium sulfate precipitation | |
| 147 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title