missing translation for 'onlineSavingsMsg'
Learn More

Lys-SMCC-DM1, MedChemExpress

Product Code. 16486407
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486407 5 mg 5mg
16490959 1 mg 1mg
16413678 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486407 Supplier Medchem Express Supplier No. HY101982

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lys-SMCC-DM1 (Lys-Nε-MCC-DM1) is a linker-payload component that has the potential to inhibit tubulin polymerization.Lys-SMCC-DM1 is the active metabolite of T-DM1. T-DM1 is a human epidermal growth factor receptor 2 (HER2)-targeting ADC with a tubulin polymerization inhibitor DM1.

Lys-SMCC-DM1 (Lys-Nε-MCC-DM1) is a linker-payload component that has the potential to inhibit tubulin polymerization.Lys-SMCC-DM1 is the active metabolite of T-DM1. T-DM1 is a human epidermal growth factor receptor 2 (HER2)-targeting ADC with a tubulin polymerization inhibitor DM1.

Specifications

Chemical Name or Material Lys-SMCC-DM1
CAS 1281816-04-3
Purity Grade Notes Research
Molecular Formula C53H75ClN6O15S
Quantity 5 mg
Synonym Lys-Nε-MCC-DM1
SMILES C[C@]1([C@@](CC(N(C(C=C2C=C3OC)=C3Cl)C)=O)([H])OC([C@H](C)N(C)C(CCS[C@@H](CC4=O)C(N4C[C@@H]5CC[C@@H](C(NCCCC[C@H](N)C(O)=O)=O)CC5)=O)=O)=O)[C@H]([C@@H]([C@](OC6=O)([H])C[C@]([C@](/C=C/C=C(C)/C2)([H])OC)(N6)O)C)O1
Molecular Weight (g/mol) 1103.71
Formula Weight 1103.71
Grade Research
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.