missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LYRM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556448
This item is not returnable.
View return policy
Description
Immunogen sequence: RAMLRESKRF SAYNYRTYAV RRIRDAFREN KNVKDPVEIQ TLVNKAKRDL GVIRRQVHIG QLYSTDKLII ENRD Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
Required for nuclear and mitochondrial iron-sulfur protein biosynthesis.
Specifications
| LYRM4 | |
| Polyclonal | |
| Unconjugated | |
| LYRM4 | |
| BC034664; C6orf149; CGI-203; COXPD19; Gm903; homolog of yeast Isd11; ISD11; LYR motif containing 4; LYR motif-containing protein 4; Lyrm4; mitochondrial matrix Nfs1 interacting protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57128 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HD34 | |
| LYRM4 | |
| Recombinant protein corresponding to Human LYRM4. Recombinant protein control fragment (Product #RP-95016). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction