missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LYRM2 Polyclonal Antibody

Product Code. 12809072
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12809072 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12809072 Supplier Invitrogen™ Supplier No. PA520921

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is A549 cell lysate. PA5-20921 can be used with blocking peptide PEP-1035.

The Lyr motif found in the LYR-motif containing protein family is similar to that found in the sacchromyces cerevisiae protein ISD11, an iron-sulfur protein in the mitochondria that is thought to play a role in iron homeostasis. No known function has been assigned to LYRM2, although LYRM1 is thought to be involved in preadipocyte progression and LYRM3 has been suggested to be a candidate gene for the branchio-oto-renal (BOR) syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYRM2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LYRM2
Gene Accession No. B2GV91, Q8R033, Q9NU23
Gene Alias 2610208E05Rik; DJ122O8.2; LYR motif containing 2; LYR motif-containing protein 2; Lyrm2; novel complex 1 (LYR family) protein
Gene Symbols LYRM2
Host Species Rabbit
Immunogen A 16 amino acid peptide near the carboxy terminus of human LYRM2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108755, 57226, 690354
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.