missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LYRM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520920
This item is not returnable.
View return policy
Description
A suggested positive control is human liver tissue lysate. PA5-20920 can be used with blocking peptide PEP-1034.
LYRM1 was one of many genes identified through a suppression subtractive hybridization comparing their expression in omental adipose tissue of obese patients compared to non-obese individuals. Further study confirms that both mRNA and protein levels of LYRM1 are higher in obese individuals. LYRM1 is widely expressed, with highest levels occurring in adipose and liver tissues. It is thought that LYRM1 promotes preadipocyte proliferation and can inhibit apoptosis of preadipocytes. Ectopic LYRM1 expression did not significantly affect adipogenesis, suggesting that LYRM1 can influence adipose tissue homeostasis by modulating the size of the preadipocyte pool.
Specifications
| LYRM1 | |
| Polyclonal | |
| Unconjugated | |
| Lyrm1 | |
| 1110065L10Rik; 2310004B22Rik; 4930404J24Rik; A211C6.1; LYR motif containing 1; LYR motif-containing protein 1; LYRM1; RGD1563498 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 365361, 57149, 73919 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| B2RYU8, O43325, Q9CQB7 | |
| Lyrm1 | |
| A 17 amino acid peptide near the center of human LYRM1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction