missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LYPLAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545807
This item is not returnable.
View return policy
Description
Peptide sequence: EETNSMLKSL GVTTKFHSFP NVYHELSKTE LDILKLWILT KLPGEMEKQK Sequence homology: Dog: 82%; Guinea Pig: 100%; Horse: 85%; Human: 100%; Rabbit: 92%; Rat: 85%.
LYPLAL1 does not exhibit phospholipase nor triacylglycerol lipase activity. It is able to hydrolyze only short chain substrates due to its shallow active site.
Specifications
| LYPLAL1 | |
| Polyclonal | |
| Unconjugated | |
| Lyplal1 | |
| BC027340; KIAA1238; LYPLAL1; lysophospholipase like 1; lysophospholipase-like 1; lysophospholipase-like protein 1; Q96AV0; Q96AVO | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 127018 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5VWZ2 | |
| Lyplal1 | |
| Synthetic peptide directed towards the C-terminal of human LYPLAL1 (aa 188-237). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction