Learn More
Invitrogen™ LYPLA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527653
Description
Recommended positive controls: 293T, A431, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (99%), Rat (99%), Chicken (88%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. There are alternatively spliced transcript variants described for this gene but the full length nature is not known yet.
Specifications
| LYPLA2 | |
| Polyclonal | |
| Unconjugated | |
| LYPLA2 | |
| Acyl-protein thioesterase 2; APT2; APT-2; DJ886K2.4; LPL-II; Lypla2; LysoII; Lysophospholipase 2; lysophospholipase II; lysoPLA II; mLyso II; testicular tissue protein Li 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11313, 26394 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O95372, Q9WTL7 | |
| LYPLA2 | |
| Recombinant protein encompassing a sequence within the center region of human LYPLA2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.