missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYPLA2 Antibody (02), Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £255.00
Specifications
| Antigen | LYPLA2 |
|---|---|
| Clone | 02 |
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670897
|
Novus Biologicals
NBP3-05819-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694928
|
Novus Biologicals
NBP3-05819-50ul |
50 μg |
£173.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LYPLA2 Monoclonal antibody specifically detects LYPLA2 in Human samples. It is validated for ELISASpecifications
| LYPLA2 | |
| ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| acyl-protein thioesterase 2, APT2, DJ886K2.4, EC 3.1.2, EC 3.1.2.-, LPL-II, lysophospholipase IIAPT-2LysoPLA II | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LYPLA2 (Uniprot#: O95372; Met 1-Val 231). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 02 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Endocrinology, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 11313 | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title