missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene hydrolyzes lysophosphatidylcholine in both monomeric and micellar forms. The use of alternate polyadenylation sites has been found for this gene. There are alternatively spliced transcript variants described for this gene but the full length nature is not known yet.
Specifications
Specifications
| Antigen | LYPLA1 |
| Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.21 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | LYPLA1 |
| Gene Accession No. | O75608, P70470, P97823 |
| Gene Alias | Acyl protein thioesterase 1, APT 1, APT1, hAPT1, LPL I, LPL1, LYPLA1, Lysophospholipase 1, lysophospholipase I, LYSOPLA, LysoPLA I |
| Gene Symbols | Lypla1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?