missing translation for 'onlineSavingsMsg'
Learn More

LYPLA1 Antibody (3D5), Novus Biologicals™

Product Code. 18369838 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369838 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369838 Supplier Novus Biologicals Supplier No. H00010434M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LYPLA1 Monoclonal antibody specifically detects LYPLA1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYPLA1
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 3D5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH08652
Gene Alias acyl-protein thioesterase-1, APT1, APT-1, EC 3.1.2, EC 3.1.2.-, hAPT1, LPL1acyl-protein thioesterase 1, LPL-I, Lysophospholipase 1, lysophospholipase ILYSOPLA, lysophospholipid-specific lysophospholipase, LysoPLA I
Host Species Mouse
Immunogen LYPLA1 (AAH08652.1, 66 a.a. ~ 151 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NVAMPSWFDIIGLSPDSQEDESGIKQAAENIKALIDQEVKNGIPSNRIILGGFSQGGALSLYTALTTQQKLAGVTALSCWLPLRAS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10434
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.