missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LYPD6B Monoclonal Antibody (634CT7.10.1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524725
This item is not returnable.
View return policy
Description
LYPD6B Monoclonal Antibody for Western Blot
Believed to act as a modulator of nicotinic acetylcholine receptors (nAChRs) activity. In vitro acts on nAChRs in a subtype- and stoichiometry-dependent manner. Modulates specifically alpha-3(3):beta-4(2) nAChRs by enhancing the sensitivity to ACh, decreasing ACh-induced maximal current response and increasing the rate of desensitization to ACh; has no effect on alpha-7 homomeric nAChRs; modulates alpha-3(2):alpha-5:beta-4(2) nAChRs in the context of CHRNA5/alpha-5 variant Asn-398 but not its wild-type sequence. [UniProt].
Specifications
| LYPD6B | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.09% sodium azide; pH 7.4 | |
| Q8NI32, Q9D7F2 | |
| LYPD6B | |
| KLH conjugated synthetic peptide between 79-108 amino acids from the Central region of human LYPD6B. | |
| RUO | |
| 130576, 71897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 634CT7.10.1 | |
| Unconjugated | |
| LYPD6B | |
| 2310010M24Rik; AW049525; cancer/testis antigen 116; CT116; LY6/PLAUR domain containing 6B; ly6/PLAUR domain-containing protein 6B; Lypd6b; LYPD7 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction