missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYPD1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | LYPD1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18351734
|
Novus Biologicals
NBP3-04851-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18352084
|
Novus Biologicals
NBP3-04851-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LYPD1 Polyclonal specifically detects LYPD1 in Mouse samples. It is validated for Western Blot.Specifications
| LYPD1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 116372 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| FLJ41033, LY6/PLAUR domain containing 1, ly6/PLAUR domain-containing protein 1, LYPDC1, MGC29643, PHTS, Putative HeLa tumor suppressor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 21-141 of human LYPD1 (NP_653187.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title