missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lyp/PTPN22 Antibody [DyLight 650], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals AF3428W
This item is not returnable.
View return policy
Description
Lyp/PTPN22 Polyclonal antibody specifically detects Lyp/PTPN22 in Human samples. It is validated for Western Blot,Immunocytochemistry
Specifications
| Lyp/PTPN22 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| DyLight 650 | |
| EC 3.1.3.48, Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP, Lymphoid phosphatase, lymphoid-specific protein tyrosine phosphatase, LYP, Lyp1, LYP2, PEP, PEST-domain phosphatase, protein tyrosine phosphatase, non-receptor type 22 (lymphoid), protein tyrosine phosphatase, non-receptor type 8, PTPN8LYP1, tyrosine-protein phosphatase non-receptor type 22 | |
| E. coli-derived recombinant human Lyp, Ser306-Ser684, Accession # Q9Y2R2 | |
| 0.1 mL | |
| Protein Phosphatase | |
| 26191 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction