missing translation for 'onlineSavingsMsg'
Learn More

Lyp/PTPN22 Antibody (4F6), Novus Biologicals™

Product Code. 18323579 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323579 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323579 Supplier Novus Biologicals Supplier No. H00026191M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lyp/PTPN22 Monoclonal antibody specifically detects Lyp/PTPN22 in Human, Mouse, Porcine samples. It is validated for Western Blot, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lyp/PTPN22
Applications Western Blot, ELISA, KnockDown
Classification Monoclonal
Clone 4F6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH17785
Gene Alias EC 3.1.3.48, Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP, Lymphoid phosphatase, lymphoid-specific protein tyrosine phosphatase, LYP, Lyp1, LYP2, PEP, PEST-domain phosphatase, protein tyrosine phosphatase, non-receptor type 22 (lymphoid), protein tyrosine phosphatase, non-receptor type 8, PTPN8LYP1, tyrosine-protein phosphatase non-receptor type 22
Host Species Mouse
Immunogen PTPN22 (AAH17785, 1 a.a. ~ 179 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDQREILQKFLDEAQSKKITKEEFANEFLKLKRQSTKYKADKTYPTTVAEKPKNIKKNRYKDILPYDYSRVELSLITSDEDSSYINANFIKGVYGPKAYIATQGPLSTTLLDFWRMIWEYSVLIIVMACMEYEMGKEAEKRKSDYIIRTLKVKFNSVSVILAHQTSLQNLFSQITPAHF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 26191
Target Species Human, Mouse, Pig
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.