missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Lyovapor™ L-300 Continuous Freeze Dryer

Product Code. 17913351 Shop All Buchi Products
Change view
Click to view available options
No. of Shelves:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Shelves
17913351 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913351 Supplier BUCHI Supplier No. 11L300PH2628112003
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Continuous L-300 Lyovapor™ is a unique freeze dryer, providing unlimited freeze drying for the first time with BUCHI's Infinite-Technology™. Dual condensers allow for continuous sublimation of both water and organic based solvents.

  • Infinite-Technology™, allowing for continuous freeze drying due to dual condensers
  • -105°C lowest condenser temperature
  • Automated cleaning
  • Sublimiation of both water and organic-based solvents
  • Advanced software for creating and managing methods, data record and reporting

Lyovapor™ L-300 Pro Freeze Dryer

  • Vacuum Pump: Edwards nXDS10ic Dry Scroll
  • Drying: Heatable shelves 4-position or 6-position, 12-position manifold cover
TRUSTED_SUSTAINABILITY

Specifications

Type Product Shelves
Capacity (Metric) >12 kg/24 hr.
No. of Shelves 6
Temperature Range (Metric) Ambient to -105°C
Material Stainless Steel
Plug Type All except North America
Voltage 220/240 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.