Learn More
BUCHI Lyovapor L-210 Freeze Dryer
GREENER_CHOICE
Product Code. 31921129 Shop All Buchi Products
Change view
Click to view available options
Model:
L-210 Basic
L-210 Basic Defrosting
L-210 Pro
L-210 Pro Defrosting
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
31921129 L-210 Basic Each
31921128 L-210 Basic Defrosting Each
31921130 L-210 Pro Each
31921132 L-210 Pro Defrosting Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31921129 Supplier BUCHI Supplier No. 11L210IN0000
Includes
The Loyvapor L-210 features groundbreaking EcoStream technology, making it the most eco-friendly laboratory freeze dryer
Requires
The Loyvapor L-210 features groundbreaking EcoStream technology, making it the most eco-friendly laboratory freeze dryer
A freeze dryer designed for efficient, continuous operation with features that support environmentally conscious lab workflows.
  • -55°C condenser suitable for aqueous and organic-based solvent mixtures
  • 6 kg ice condensing capacity for standard laboratory applications
  • EcoStream technology uses natural coolants
  • Global Warming Potential (GWP) of only 4
  • Reduced heat output and noise emissions in the lab
  • Infinite-Control technology offers process control via display, software, and mobile app
  • Sample protection mode prevents sample collapse
Order Info
freeze dryer, lyophilizer
Certifications
The Loyvapor L-210 features groundbreaking EcoStream technology, making it the most eco-friendly laboratory freeze dryer
Compliance
The Loyvapor L-210 features groundbreaking EcoStream technology, making it the most eco-friendly laboratory freeze dryer
Warranty
freeze dryer, lyophilizer
The Loyvapor L-210 features groundbreaking EcoStream technology, making it the most eco-friendly laboratory freeze dryer
TRUSTED_SUSTAINABILITY
Specifications
Type Freeze Dryer
Capacity (Metric) 6 kg ice
No. of Shelves 0
Temperature Range (Metric) -55°C
Model L-210 Basic
Material Stainless Steel
Plug Type North America
Voltage 220/240 V