missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Lyovapor™ L-200

Freeze dryer with Infinite-Control™

Brand:  BUCHI 11L200IN1422003001


Additional Details : Weight : 145.00000kg

 View more versions of this product

Product Code. 17224724

  • £18745.00 / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

  • High quality freeze drying (-55°C, 6 kg)
  • High level of automation
  • Infinite Control™ includes:
    • Easy method creation
    • Method running
    • Data logging
    • Chart recording in real-time and interruption if needed
Specifications

Specifications

6 kg
4
Stainless Steel
Vacuum pump, Drying chamber manifold
23.15 in.
13.2 lb.
-55°C to +30°C
Edwards nXDS6iC
20.1 in.
240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers