missing translation for 'onlineSavingsMsg'
Learn More

Lyn-IN-1, MedChemExpress

Product Code. 16495716
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495716

Brand: Medchem Express HY12039

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Lyn-IN-1 (Bafetinib analog) is a potent and selective dual Bcr-Abl/Lyn inhibitor, extracted from patent WO2014169128A1.

Lyn-IN-1 (Bafetinib analog) is a potent and selective dual Bcr-Abl/Lyn inhibitor, extracted from patent WO2014169128A1.

Specifications

Chemical Name or Material Lyn-IN-1
Physical Form Solid
CAS 887650-05-7
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C30H31F3N8O
For Use With (Application) Cancer-Kinase/protease
Synonym Bafetinib analog
Solubility Information DMSO : 50 mg/mL (86.71 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C)C(NC2=NC=C(C3=CN=CN=C3)C=N2)=C1)C4=CC=C(CN5C[C@@H](N(C)C)CC5)C(C(F)(F)F)=C4
Molecular Weight (g/mol) 576.62
Formula Weight 576.62
Percent Purity 98.66%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.