missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Lyn Human Protein, Active

Product Code. p-7228102
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17300274

Brand: Eurofins DiscoverX 14510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal his6-tagged recombinant full-length human Lyn, for use in Kinase Assays.

The Lyn protein is a member of the Src family of non-receptor-associated protein-tyrosine kinases. LYN A differs from LYN B in having an additional 21 amino acids at the amino terminus. Lyn undergoes a concentration-dependent autophosphorylation, thus activating the catalytic site and decreasing the accessibility of the SH2 domain. Lyn phosphorylates and activates Tec, non-receptor tyrosine kinase that is coexpressed with Lyn and acts downstream of Lyn in intracellular signaling pathways. Lyn tyrosine kinase has been shown to be necessary for IgE-receptor (FceRI)-mediated mast-cell activation.

EMBL M16038

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 59.6 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade ≥75%
Protein Lyn, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.