missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Lyn Isoform B, His Tag Recombinant Protein

Product Code. 15468383
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 15468383

Brand: Invitrogen™ PR3725B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

LYN is a SRC family kinase expressed predominantly in hematopoietic cells. Activation of LYN has been shown to lead to imatinib resistance in CML patients. LYNB and the slightly shorter LYNA are the two isoforms of LYN. Diseases associated with LYN include Sarcoma and Myeloid Leukemia. Among its related pathways are Signaling by Erythropoietin and Regulation of actin dynamics for phagocytic cup formation. Gene Ontology (GO) annotations related to Lyn Isoform B include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of Lyn Isoform B is HCK.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07948
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.05% NP-40, 0.05 mM EDTA, 100 mM NaCl, 1 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 4067
Molecular Weight (g/mol) 57.4 kDa
Name Human Lyn Isoform B, His Tag
Quantity 100 μg
Regulatory Status RUO - research use only
Gene Alias LYN
Common Name Lyn Isoform B
KinaseFamily SRC Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.