missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lyn Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-93453
This item is not returnable.
View return policy
Description
Lyn Polyclonal specifically detects Lyn in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Lyn | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| EC 2.7.10, EC 2.7.10.2, FLJ26625, JTK8, tyrosine-protein kinase Lyn, v-yes-1 Yamaguchi sarcoma viral related oncogene homolog, Yamaguchi sarcoma viral (v-yes-1) related oncogene homolog | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Cow and Mouse. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_002341.1 | |
| LYN | |
| Peptide with sequence C-KTQPVRNTERT corresponding to internal region according to NP_002341.1. | |
| 0.1 mg | |
| Protein Kinase, Signal Transduction | |
| 4067 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction