missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Lyn Monoclonal antibody specifically detects Lyn in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | Lyn |
| Applications | Western Blot, Immunocytochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | LYN-01 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.7.10, EC 2.7.10.2, FLJ26625, JTK8, tyrosine-protein kinase Lyn, v-yes-1 Yamaguchi sarcoma viral related oncogene homolog, Yamaguchi sarcoma viral (v-yes-1) related oncogene homolog |
| Host Species | Mouse |
| Immunogen | Bacterially expressed recombinant fragment of human Lyn, aa 8 - 238 (NM_001111097.2). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?