missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lyn Antibody (LYN-01), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-519AF405
This item is not returnable.
View return policy
Description
Lyn Monoclonal specifically detects Lyn in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Lyn | |
| Monoclonal | |
| Alexa Fluor 405 | |
| P07948 | |
| LYN | |
| Bacterially expressed recombinant fragment of human Lyn, aa 8 - 238 (NM_001111097.2). | |
| 0.1 mL | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 4067 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| LYN-01 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.10, EC 2.7.10.2, FLJ26625, JTK8, tyrosine-protein kinase Lyn, v-yes-1 Yamaguchi sarcoma viral related oncogene homolog, Yamaguchi sarcoma viral (v-yes-1) related oncogene homolog | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The LYN-01 reacts with Lyn (p56/p53), a non-receptor Src-family tyrosine kinase expressed in hematopoietic tissues. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction