missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Lyn Monoclonal antibody specifically detects Lyn in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot
Specifications
Specifications
| Antigen | Lyn |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | JE44-75 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:1000 |
| Formulation | 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol |
| Gene Alias | EC 2.7.10, EC 2.7.10.2, FLJ26625, JTK8, tyrosine-protein kinase Lyn, v-yes-1 Yamaguchi sarcoma viral related oncogene homolog, Yamaguchi sarcoma viral (v-yes-1) related oncogene homolog |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?