missing translation for 'onlineSavingsMsg'
Learn More

Lyn Antibody (338716) [FITC], Novus Biologicals™

Product Code. 30731936 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731936 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731936 Supplier Novus Biologicals Supplier No. FAB3206F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lyn Monoclonal antibody specifically detects Lyn in Human,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lyn
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 338716
Conjugate FITC
Formulation PBS
Gene Alias EC 2.7.10, EC 2.7.10.2, FLJ26625, JTK8, tyrosine-protein kinase Lyn, v-yes-1 Yamaguchi sarcoma viral related oncogene homolog, Yamaguchi sarcoma viral (v-yes-1) related oncogene homolog
Host Species Mouse
Immunogen E. coli-derived recombinant human Lyn, Met1-Ile67, Accession # P07948
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4067
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.