missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LYN A, GST Tag Recombinant Protein
Human LYN A, GST Tag Recombinant Protein
£402.00 - £5620.00
Specifications
Accession Number | NP_002341.1 |
---|---|
Gene Alias | JTK8, FLJ26625 |
KinaseFamily | SRC Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15499053
|
Invitrogen™
PV6448 |
10μg |
£402.00
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15409063
|
Invitrogen™
PV6449 |
100μg |
£1306.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15419063
|
Invitrogen™
PV6450 |
1mg |
£5620.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
LYN is a protein-coding gene and is associated with diseases such as alexia and chorea-acanthocytosis. Activation of LYN has been shown to lead to imatinib resistance in CML patients. LYN B and the slightly shorter LYN A are the two isoforms of LYN. An important paralog of this gene is FRK. The full length LYN A protein enables researchers to study important mechanisms underlying disease.
- Increase disease relevance by interrogating a purified and active full-length LYN A reagent
- Uncover novel compounds or hits specific to the full length form
- Advance your research using the full-length protein and validated assays
- Assay Platforms: LanthaScreen™ TR-FRET Kinase Activity Assay, LanthaScreen™ Eu Kinase Binding Assay, and Z’-LYTE™ Kinase Activity Assay
Specifications
NP_002341.1 | |
SRC Kinase Family | |
Recombinant |
JTK8, FLJ26625 | |
Human | |
GST Tag |