missing translation for 'onlineSavingsMsg'
Learn More

Lymphotoxin-alpha/TNF-beta Antibody (3F12.2D3), PerCP, Novus Biologicals™

Product Code. 18333091 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333091 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333091 Supplier Novus Biologicals Supplier No. NBP281105PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Lymphotoxin-alpha/TNF-beta Monoclonal specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lymphotoxin-alpha/TNF-beta
Applications Flow Cytometry
Classification Monoclonal
Clone 3F12.2D3
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias LT, LT-alpha, lymphotoxin alpha (TNF superfamily, member 1), TNFBlymphotoxin-alpha, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1
Gene Symbols LTA
Host Species Rabbit
Immunogen 3F12 was prepared by hyperimmunizing BALB/c mice with purified recombinant human LTalpha expressed in E. coli - the hybridoma clone was selected for its ability to bind LTalpha3 by ELISA. The CDRs of the antibody were grafted to the framework regions of the mice mAb 2G7 to create the chimeric version of the antibody.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 4049
Test Specificity 3F12 binds to human LTalpha3 and LT-alpha-beta (with a KD of ∽0.3 nM, determined by BIACORE; ∽ 37 pM, determined by ELISA) in both its soluble homotrimeric and membrane heterotrimeric forms. LTalpha belongs to the tumor necrosis factor (TNF) superfamily and is secreted as a homo-trimer (LTalpha3), or is expressed on the cell surface in complex with LTbeta. Lymphotoxin is produced by lymphocytes and in its homotrimeric form binds to TNFRSF1A/TNFR1, TNFRSF1B/TNFBR and TNFRSF14/HVEM. LTalpha3 signaling induces target cells to upregulate many chemokines and cytokines in an NFkB-dependent manner.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.