missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lymphotoxin-alpha/TNF-beta Antibody (074), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89370AF594
This item is not returnable.
View return policy
Description
Lymphotoxin-alpha/TNF-beta Monoclonal specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Sandwich ELISA Detection.
Specifications
| Lymphotoxin-alpha/TNF-beta | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human TNFa Mouse TNFa Human TNFRII | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 74 | |
| Sandwich ELISA Detection | |
| LT, LT-alpha, lymphotoxin alpha (TNF superfamily, member 1), TNFBlymphotoxin-alpha, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Lymphotoxin-alpha/TNF-beta (P01374; Leu35-Leu205). | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
| 4049 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction