missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lymphotactin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA122497
This item is not returnable.
View return policy
Description
PA1-22497 detects Lymphotactin in Mouse samples.
Lymphotactin, otherwise known as XCL1, is a member of the C-chemokine family that uniquely acts as a chemoattractant for lymphocytes, but not monocytes or neutrophils, suggesting an important regulatory role in lymphocyte trafficking and inflammation. Lymphotactin, shown to signal through chemokine XC receptor 1 (XCR1), is detected at the highest level in the spleen and at low levels in the lungs, small intestine and colon, and is abundantly and selectively expressed by activated T cells, in particular CD8+ T cells, and is also produced by NK cells.
Specifications
| Lymphotactin | |
| Polyclonal | |
| Unconjugated | |
| Xcl1 | |
| AI661682; ATAC; C motif chemokine 1; chemokine (C motif) ligand 1; cytokine SCM-1; Lptn; LTN; Lymphotactin; lymphotaxin; SCM1; SCM-1; SCM1A; SCM-1a; SCM1alpha; SCM-1alpha; SCM-1-alpha; SCM1beta; SCM-1beta; SCM-1-beta; SCYC1; single cysteine motif 1a; small inducible cytokine subfamily C, member 1 (lymphotactin); small-inducible cytokine C1; XC chemokine ligand 1; X-C motif chemokine ligand 1; XCL1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16963 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P47993 | |
| Xcl1 | |
| Recombinant fragment, corresponding to amino acids 1-92 of Mouse Lymphotactin. | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction