missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LYL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567370
This item is not returnable.
View return policy
Description
Immunogen sequence: CPPQAQAEVGP TMTEKAEMVC A Highest antigen sequence identity to the following orthologs - mouse 82%, rat 82%.
Lyl-1, TAL1 and TAL2 are part of a family of basic helix-loop-helix (bHLH) proteins implicated in T cell acute leukemia. TAL1, also designated SCL, is a serine phosphoprotein and basic helix-loop-helix transcription factor known to regulate embryonic hematopoiesis. TAL2 is a protein involved in T cell acute lymphoblastic leukemia through a chromosomal translocation involving TAL2 and T cell receptor β chain genes. TAL2 includes a helix-loop-helix protein dimerization and DNA-binding domain that is homologous to TAL1 and Lyl-1 proto-oncogenes. Lyl-1 (lymphoblastic leukemia-derived sequence 1) is a nuclear protein. Endogenous Lyl-1 exists in complex with E2α proteins. Lyl-1 and E2α protein can form heterodimeric complexes with distinctive DNA-binding properties in hematolymphoid cells. Lyl-1 is involved in a chromosomal aberration which causes a form of T cell acute lymphoblastic leukemia (T-ALL).
Specifications
| LYL1 | |
| Polyclonal | |
| Unconjugated | |
| LYL1 | |
| bHLHa18; Class A basic helix-loop-helix protein 18; LYL1; Lyl-1; LYL1, basic helix-loop-helix family member; lymphoblastic leukemia associated hematopoiesis regulator 1; lymphoblastic leukemia derived sequence 1; lymphoblastic leukemia-derived sequence 1; lymphoblastomic leukemia 1; protein lyl-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4066 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P12980 | |
| LYL1 | |
| Recombinant Human LYL1. Recombinant protein control fragment (Product #RP-108028). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction