missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LYL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568772
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Mouse: 100%; Rat: 86%.
Lyl-1, TAL1 and TAL2 are part of a family of basic helix-loop-helix (bHLH) proteins implicated in T cell acute leukemia. TAL1, also designated SCL, is a serine phosphoprotein and basic helix-loop-helix transcription factor known to regulate embryonic hematopoiesis. TAL2 is a protein involved in T cell acute lymphoblastic leukemia through a chromosomal translocation involving TAL2 and T cell receptor β chain genes. TAL2 includes a helix-loop-helix protein dimerization and DNA-binding domain that is homologous to TAL1 and Lyl-1 proto-oncogenes. Lyl-1 (lymphoblastic leukemia-derived sequence 1) is a nuclear protein. Endogenous Lyl-1 exists in complex with E2α proteins. Lyl-1 and E2α protein can form heterodimeric complexes with distinctive DNA-binding properties in hematolymphoid cells. Lyl-1 is involved in a chromosomal aberration which causes a form of T cell acute lymphoblastic leukemia (T-ALL).
Specifications
| LYL1 | |
| Polyclonal | |
| Unconjugated | |
| LYL1 | |
| bHLHa18; Class A basic helix-loop-helix protein 18; LYL1; Lyl-1; LYL1, basic helix-loop-helix family member; lymphoblastic leukemia associated hematopoiesis regulator 1; lymphoblastic leukemia derived sequence 1; lymphoblastic leukemia-derived sequence 1; lymphoblastomic leukemia 1; protein lyl-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 304663 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q66HH3 | |
| LYL1 | |
| synthetic peptide directed towards the C-terminal region of rat LYL1. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction