missing translation for 'onlineSavingsMsg'
Learn More

Lycorine Hydrochloride, TRC

Product Code. 18061930
Change view
Click to view available options
Quantity:
20 mg
50 mg
500 mg
Unit Size:
20mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18061930 500 mg 500mg
18052740 50 mg 50mg
18090320 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18061930 Supplier Toronto Research Chemicals Supplier No. TRCL487550500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2188-68-3
Molecular Formula C16 H17 N O4 . Cl H
Molecular Weight (g/mol) 323.7714
Synonym Licorin Hydrochloride, 3,3a-Didehydrolycoran-1α,2β-diol Hydrochloride, (1α,2β)- 3,12-Didehydro-9,10-[methylenebis(oxy)]-galanthan-1,2-diol, 2,4,5,7,12b,12c-Hexahydro-1H-[1,3]dioxolo[4,5-j]pyrrolo[3,2,1-de]phenanthridine-1,2-diol Hydrochloride, (1S,2S,12bS,12cS)-2,4,5,7,12b,12c-Hexahydro-1H-[1,3]dioxolo[4,5-j]pyrrolo[3,2,1-de]phenanthridine-1,2-diol
SMILES Cl.O[C@H]1C=C2CCN3Cc4cc5OCOc5cc4[C@H]([C@@H]1O)[C@@H]23

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H17NO4.ClH/c18-11-3-8-1-2-17-6-9-4-12-13(21-7-20-12)5-10(9)14(15(8)17)16(11)19;/h3-5,11,14-16,18-19H,1-2,6-7H2;1H/t11-,14-,15+,16+;/m0./s1
Formula Weight 323.0924
Chemical Name or Material Lycorine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.