missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Lycorine hydrochloride

Product Code. 16464807
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464807 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16464807 Supplier MedChemExpress Supplier No. HYN0289
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lycorine hydrochloride is the main active ingredient of the herbal medicine derived from Lycoris radia and is also a melanoma vasculogenic inhibitor and has anti-tumor activity. Lycorine hydrochloride effectively inhibits mitotic proliferation of Hey1B cells (IC50 of 1.2 μM).

Specifications

Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 323.77 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 2188-68-3
Product Type Natural Products
Purity 99.35%
Solubility DMSO : 25 mg/mL (77.22 mM∣Need ultrasonic) ∣H2O : 16.67 mg/mL (51.49 mM∣Need ultrasonic)
Target Autophagy∣Bacterial∣Virus Protease
Quantity 50 mg
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.